N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0505-25G F0505-1G F0505-5G
TCF0505-25GEA 104.77 USD
TCF0505-25G TCF0505-1G TCF0505-5G
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine ≥98.0% (by HPLC, titration analysis)
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine

CAS Number: 71989-35-0
Molecular Formula: C23H27NO5
Molecular Weight: 397.47
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 133
Specific rotation [a]20/D: 15 deg (C=1, EtOAc)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: O-tert-Butyl-Nalpha-Fmoc-L-threonine, Nalpha-Fmoc-O-tert-butyl-L-threonine, Fmoc-Thr(tBu)-OH

Formula: C₂₃H₂₇NO₅
Melting Pt: 133 °C
MDL Number: MFCD00077075
CAS Number: 71989-35-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b7cac1af38ae52c71aef54cb89efea73ee232ce&vid=8b7cac1af38ae52c71aef54cb89efea73ee232ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T,HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR