5-Hexenyl acetate ≥98.0%
Supplier: TCI America
A1338-25ML
A1338-500ML
TCA1338-025MLEA
60
USD
TCA1338-025ML
TCA1338-500ML
5-Hexenyl acetate ≥98.0%
5-Hexenyl acetate
CAS Number: 5048-26-0
MDL Number: MFCD00060856
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 61
Specific Gravity (20/20): 0.90
Other Product Names / Synonyms: 6-Acetoxy-1-hexene, Acetic Acid 5-Hexenyl Ester
Formula:
C₈H₁₄O₂ MW: 142.20 g/mol Boiling Pt: 172 °C Density: 0.90 (20 °C) Flash Pt: 61 °C |
MDL Number:
MFCD00060856 CAS Number: 5048-26-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...