5-Formylsalicylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0400-25G F0400-5G
TCF0400-25GEA 88.9 USD
TCF0400-25G TCF0400-005G
5-Formylsalicylic acid ≥98.0% (by HPLC, titration analysis)
5-Formylsalicylic acid

CAS Number: 616-76-2
MDL Number: MFCD00006945
Molecular Formula: C8H6O4
Molecular Weight: 166.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 250


Other Product Names / Synonyms: 5-Formyl-2-hydroxybenzoic Acid

Formula: C₈H₆O₄
Melting Pt: 250 °C
MDL Number: MFCD00006945
CAS Number: 616-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edd23d1ecccd7de9781b23ba7d11d67ae38d87f3&vid=edd23d1ecccd7de9781b23ba7d11d67ae38d87f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR