4-Fluorophenyl chlorothionoformate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: O-(4-Fluorophenyl) Chlorothioformate, Chlorothioformic Acid O-(4-Fluorophenyl) Ester

F0365-1G F0365-5G
TCF0365-001GEA 52.3 USD
TCF0365-001G TCF0365-005G
4-Fluorophenyl chlorothionoformate ≥98.0% (by titrimetric analysis)
4-Fluorophenyl chlorothionoformate

CAS Number: 42908-73-6
MDL Number: MFCD00134402
Molecular Formula: C7H4ClFOS
Molecular Weight: 190.62
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 84
Specific Gravity (20/20): 1.37


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Chlorothioformic Acid O-(4-Fluorophenyl) Ester, O-(4-Fluorophenyl) Chlorothioformate

Formula: C₇H₄ClFOS
MW: 190.62 g/mol
Boiling Pt: 82 °C
Density: 1.37 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00134402
CAS Number: 42908-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f81e1a37ea092997b48cadb6fc98705d4e3351d&vid=6f81e1a37ea092997b48cadb6fc98705d4e3351d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR