2-Phenylethyl formate

Supplier: TCI America

F0363-25G F0363-500G
TCF0363-025GEA 33 USD
TCF0363-025G TCF0363-500G
2-Phenylethyl formate
2-Phenylethyl formate

CAS Number: 104-62-1
MDL Number: MFCD00021046
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 117
Flash Point (°C): 103
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Formic Acid Phenethyl Ester, Phenethyl Formate, Formic Acid 2-Phenylethyl Ester

Formula: C₉H₁₀O₂
Boiling Pt: 117 °C
Density: 1.06 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00021046
CAS Number: 104-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b7c7cc41ff91df396940af3c22a9d133532576f&vid=0b7c7cc41ff91df396940af3c22a9d133532576f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR