3-Furanmethanol ≥98.0%

Supplier: TCI America

Synonyms: 3-Furyl Alcohol, 3-(Hydroxymethyl)furan

F0360-10G F0360-25G
TCF0360-010GEA 274.1 USD
TCF0360-010G TCF0360-25G
3-Furanmethanol ≥98.0%
3-Furanmethanol

CAS Number: 4412-91-3
MDL Number: MFCD00005352
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 38
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: 3-Furyl Alcohol, 3-(Hydroxymethyl)furan

Formula: C₅H₆O₂
MW: 98.10 g/mol
Boiling Pt: 100 °C
Density: 1.14 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00005352
CAS Number: 4412-91-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74d57504d7759fb3a7f0e03223bd1eba3f9ac225&vid=74d57504d7759fb3a7f0e03223bd1eba3f9ac225&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR