Methyl-2,2-difluoro-2-(fluorosulfonyl)acetate ≥97.0%
Supplier: TCI America
F0311-5G
F0311-25G
TCF0311-005GEA
37.5
USD
TCF0311-005G
TCF0311-025G
Methyl-2,2-difluoro-2-(fluorosulfonyl)acetate ≥97.0%
Methyl 2,2-difluoro-2-(fluorosulphonyl)acetate
CAS Number: 680-15-9
MDL Number: MFCD00144316
Molecular Formula: C3H3F3O4S
Molecular Weight: 192.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 47
Specific Gravity (20/20): 1.52
Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds
Other Product Names / Synonyms: Difluoro(fluorosulfonyl)acetic Acid Methyl Ester, Methyl (Fluorosulfonyl)difluoroacetate
Formula:
C₃H₃F₃O₄S MW: 192.11 Boiling Pt: 118 °C Density: 1.52 (20 °C) Flash Pt: 47 °C |
MDL Number:
MFCD00144316 CAS Number: 680-15-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...