Furosemide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-(Aminosulphonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 4-Chloro-N-furfuryl-5-sulphamoylanthranilic acid

F0182-5G F0182-25G
TCF0182-005GEA 87.71 USD
TCF0182-005G TCF0182-025G
Furosemide ≥99.0% (by titrimetric analysis)
Furosemide

CAS Number: 54-31-9
MDL Number: MFCD00010549
Molecular Formula: C12H11ClN2O5S
Molecular Weight: 330.74
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 205


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 5-(Aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic Acid, 4-Chloro-N-furfuryl-5-sulfamoylanthranilic Acid

Formula: C₁₂H₁₁ClN₂O₅S
MW: 330.74 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00010549
CAS Number: 54-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aeedaa1708089dac6646e9cc50e908486f354792&vid=aeedaa1708089dac6646e9cc50e908486f354792&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR