(Ferrocenylmethyl)trimethylammonium bromide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

F0166-5G F0166-1G
TCF0166-5GEA 999 USD
TCF0166-5G TCF0166-001G
(Ferrocenylmethyl)trimethylammonium bromide ≥95.0% (by titrimetric analysis)
(Ferrocenylmethyl)trimethylammonium bromide

CAS Number: 106157-30-6 MDL Number: MFCD00059981 Molecular Formula: C14H20BrFeN Molecular Weight: 338.07 Purity/Analysis Method: 95.0% (T) Form: Crystal

Formula: C₁₄H₂₀BrFeN
MW: 338.07 g/mol
MDL Number: MFCD00059981
CAS Number: 106157-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e98e68abadf2798eb68c6c16fd92c4bbb54191fd&vid=e98e68abadf2798eb68c6c16fd92c4bbb54191fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR