2'-Amino-3'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-(2-Amino-3-hydroxyphenyl)ethanone
A1321-1G
A1321-5G
TCA1321-001GEA
81.6
USD
TCA1321-001G
TCA1321-5G
2'-Amino-3'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
2'-Amino-3'-hydroxyacetophenone
CAS Number: 4502-10-7
MDL Number: MFCD00191301
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 182
Formula:
C₈H₉NO₂ MW: 151.1626 g/mol Melting Pt: 182 °C |
MDL Number:
MFCD00191301 CAS Number: 4502-10-7 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...