2-Furaldehyde ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
2-Furfurol, Furfural
F0073-25G
F0073-500G
TCF0073-025GEA
26.46
USD
TCF0073-025G
TCF0073-500G
2-Furaldehyde ≥98.0%
2-Furaldehyde
CAS Number: 98-01-1
MDL Number: MFCD00003229
Molecular Formula: C5H4O2
Molecular Weight: 96.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Melting point (°C): -37
Flash Point (°C): 59
Specific Gravity (20/20): 1.16
Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: 2-Furaldehyde
Formula:
C₅H₄O₂ MW: 96.09 g/mol Boiling Pt: 160 °C Melting Pt: -37 °C Density: 1.16 (20 °C) Flash Pt: 59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003229 CAS Number: 98-01-1 EINECS: 202-627-7 UN: 1199 ADR: 6.1,II Merck Index: 13,04325 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...