Isoamyl formate ≥95.0%

Supplier: TCI America

F0054-25ML F0054-500ML
TCF0054-025MLEA 17 USD
TCF0054-025ML TCF0054-500ML
Isoamyl formate ≥95.0%
Isoamyl formate

CAS Number: 110-45-2
MDL Number: MFCD00021049
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 25
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Formic Acid Isoamyl Ester

Formula: C₆H₁₂O₂
Boiling Pt: 124 °C
Density: 0.88 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00021049
CAS Number: 110-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3b6517ca9b902f00b55a3a05295712c5fd9c3a9&vid=a3b6517ca9b902f00b55a3a05295712c5fd9c3a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR