Pentyl formate ≥95.0%

Supplier: TCI America
Danger

F0049-25ML F0049-500ML
TCF0049-025MLEA 29 USD
TCF0049-025ML TCF0049-500ML
Pentyl formate ≥95.0%
Pentyl formate

CAS Number: 638-49-3
MDL Number: MFCD00040440
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 32
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Formic Acid Amyl Ester, Pentyl Formate

Formula: C₆H₁₂O₂
Boiling Pt: 130 °C
Melting Pt: –73 °C
Density: 0.89 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00040440
CAS Number: 638-49-3
EINECS: 211-340-6
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=776e857046ad692a1cc144e1f8188b6d1d49a97b&vid=776e857046ad692a1cc144e1f8188b6d1d49a97b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR