5-Fluoro-8-quinolinol ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Fluoro-8-hydroxyquinoline

F0038-1G F0038-5G
TCF0038-001GEA 1002.2 USD
TCF0038-001G TCF0038-005G
5-Fluoro-8-quinolinol ≥99.0% (by GC, titration analysis)
5-Fluoro-8-quinolinol

CAS Number: 387-97-3
MDL Number: MFCD00059754
Molecular Formula: C9H6FNO
Molecular Weight: 163.15
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 5-Fluorooxine

Formula: C₉H₆FNO
MW: 163.15 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00059754
CAS Number: 387-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ed779591755e277823319bcf6be0ac04952c770&vid=3ed779591755e277823319bcf6be0ac04952c770&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR