7-Ethyl-3-indoleethanol ≥98.0%

Supplier: TCI America

E0824-25G E0824-5G
TCE0824-25GEA 446 USD
TCE0824-25G TCE0824-5G
7-Ethyl-3-indoleethanol ≥98.0%
7-Ethyl-3-indoleethanol

CAS Number: 41340-36-7
MDL Number: MFCD01718805
Molecular Formula: C12H15NO
Molecular Weight: 189.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 50


Other Product Names / Synonyms: 7-Ethyl-3-(2-hydroxyethyl)indole, 7-Ethyltryptophol

Formula: C₁₂H₁₅NO
Melting Pt: 50 °C
MDL Number: MFCD01718805
CAS Number: 41340-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dee6ae1b9d40f4808746fac452fbbee6325a1b0&vid=9dee6ae1b9d40f4808746fac452fbbee6325a1b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR