3-Acetyl-2-(methylaminosulfonyl)thiophene ≥98.0%

Supplier: TCI America

Synonyms: 3-Acetyl-2-thiophene-N-methylsulphonamide

A1315-5G A1315-1G
TCA1315-005GEA 552.4 USD
TCA1315-005G TCA1315-1G
3-Acetyl-2-(methylaminosulfonyl)thiophene ≥98.0%
3-Acetyl-2-(methylaminosulfonyl)thiophene

CAS Number: 138891-01-7
MDL Number: MFCD00191372
Molecular Formula: C7H9NO3S2
Molecular Weight: 219.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 105


Other Product Names / Synonyms: 3-Acetyl-2-thiophene-N-methylsulfonamide

Formula: C₇H₉NO₃S₂
MW: 219.27 g/mol
Melting Pt: 105 °C
MDL Number: MFCD00191372
CAS Number: 138891-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e729657b734a9000c34874834bd7d0ccacc125a1&vid=e729657b734a9000c34874834bd7d0ccacc125a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR