Ethyl-2,3-butadienoate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl buta-2,3-dienoate, 2,3-Butadienoic Acid Ethyl Ester

E0820-1G E0820-5G
TCE0820-1GEA 93 USD
TCE0820-1G TCE0820-5G
Ethyl-2,3-butadienoate ≥95.0%
Ethyl-2,3-butadienoate

CAS Number: 14369-81-4
MDL Number: MFCD01863567
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 47
Specific Gravity (20/20): 0.97
Lambda max.: 209 nm (MeOH)Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 2,3-Butadienoic Acid Ethyl Ester

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 80 °C
Density: 0.97 (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD01863567
CAS Number: 14369-81-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bdd3bbfb6f65b1fd8515b3d5dd9a257af9c4099&vid=2bdd3bbfb6f65b1fd8515b3d5dd9a257af9c4099&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR