3,4-Epoxytetrahydrofuran ≥97.0%

Supplier: TCI America

E0795-25G E0795-5G
TCE0795-25GEA 301 USD
TCE0795-25G TCE0795-5G
3,4-Epoxytetrahydrofuran ≥97.0%
3,4-Epoxytetrahydrofuran

CAS Number: 285-69-8
MDL Number: MFCD00800639
Molecular Formula: C4H6O2
Molecular Weight: 86.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 38
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: 3,6-Dioxabicyclo[3.1.0]hexane

Formula: C₄H₆O₂
Boiling Pt: 144 °C
Density: 1.20 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00800639
CAS Number: 285-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dbd71709746693d84a180e18e2a985b0c8b5d7b&vid=4dbd71709746693d84a180e18e2a985b0c8b5d7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR