Ethyl-3-bromo-2-(bromomethyl)propionate ≥97.0%
Supplier: TCI America
Synonyms:
Ethyl 3-bromo-2-(bromomethyl)propanoate, 3-Bromo-2-(bromomethyl)propionic Acid Ethyl Ester
E0791-25G
E0791-5G
TCE0791-25GEA
444
USD
TCE0791-25G
TCE0791-5G
Ethyl-3-bromo-2-(bromomethyl)propionate ≥97.0%
Ethyl-3-bromo-2-(bromomethyl)propionate
CAS Number: 58539-11-0
MDL Number: MFCD00034154
Molecular Formula: C6H10Br2O2
Molecular Weight: 273.95
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 113
Specific Gravity (20/20): 1.72
Other Product Names / Synonyms: 3-Bromo-2-(bromomethyl)propionic Acid Ethyl Ester
Formula:
C₆H₁₀Br₂O₂ MW: 273.95 g/mol Boiling Pt: 86 °C Density: 1.72 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00034154 CAS Number: 58539-11-0 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...