Enrofloxacin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid, 1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS Number: 93106-60-6
MDL Number: MFCD00792463
Molecular Formula: C19H22FN3O3
Molecular Weight: 359.40
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 225
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
Formula:
C₁₉H₂₂FN₃O₃ MW: 359.40 g/mol Melting Pt: 225 °C |
MDL Number:
MFCD00792463 CAS Number: 93106-60-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...