4-Ethylbenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
E0773-25G
E0773-5G
TCE0773-25GEA
148
USD
TCE0773-25G
TCE0773-5G
4-Ethylbenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
4-Ethylbenzenesulfonamide
CAS Number: 138-38-5
MDL Number: MFCD01365828
Molecular Formula: C8H11NO2S
Molecular Weight: 185.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 111
Formula:
C₈H₁₁NO₂S MW: 185.24 g/mol Melting Pt: 111 °C |
MDL Number:
MFCD01365828 CAS Number: 138-38-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...