trans-4-Ethylcyclohexanol ≥96.0% (by GC)
Supplier: TCI America
E0768-5G
E0768-1G
TCE0768-5GEA
391.3
USD
TCE0768-5G
TCE0768-1G
trans-4-Ethylcyclohexanol ≥96.0% (by GC)
trans-4-Ethylcyclohexanol
CAS Number: 19781-62-5
MDL Number: MFCD00070694
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Specific Gravity (20/20): 0.91
Formula:
C₈H₁₆O Boiling Pt: 84 °C Density: 0.91 (20 °C) |
MDL Number:
MFCD00070694 CAS Number: 19781-62-5 |
Specification Test Results
Appearance | Colourless to Almost Colourless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...