Enoxacin sesquihydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-Ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid sesquihydrate, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic Acid

E0762-5G E0762-25G
TCE0762-5GEA 197 USD
TCE0762-5G TCE0762-25G
Enoxacin sesquihydrate ≥98.0% (by HPLC, titration analysis)
Enoxacin sesquihydrate

CAS Number: 84294-96-2
Molecular Formula: C15H17FN4O3
Molecular Weight: 320.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 226


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic Acid

Formula: C₃₀H₄₀F₂N₈O₉
MW: 694.68 g/mol
Melting Pt: 226 °C
MDL Number: MFCD01747755
CAS Number: 84294-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23d93f5f03b59d04cff43c853d2a6ba658058ec9&vid=23d93f5f03b59d04cff43c853d2a6ba658058ec9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR