(-)-Epigallocatechin gallate hydrate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
EGCG hydrate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate hydrate, (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl gallate hydrate
CAS Number: 989-51-5
MDL Number: MFCD00075940
Molecular Formula: C22H18O11
Molecular Weight: 458.38
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 212
Specific rotation [a]20/D: -176 deg (C=1, EtOH)
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Regenerative Medicine Research Reagents, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals), Cancer Research Reagents, Epige
Other Product Names / Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, EGCG
Formula:
C₂₂H₁₈O₁₁ MW: 458.38 g/mol Melting Pt: 212 °C |
MDL Number:
MFCD00075940 CAS Number: 989-51-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...