(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-(1-Ethylpyrrolidin-2-yl)methanamine, (S)-2-Aminomethyl-1-ethylpyrrolidine, (S)-1-ethyl-2-aminomethylpyrrolidine
A1301-1G
A1301-5G
TCA1301-001GEA
130.8
USD
TCA1301-001G
TCA1301-5G
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine ≥98.0% (by GC, titration analysis)
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine
CAS Number: 22795-99-9
MDL Number: MFCD00191371
Molecular Formula: C7H16N2
Molecular Weight: 128.22
Purity/Analysis Method:
Formula:
C₇H₁₆N₂ MW: 128.22 g/mol Density: 0.90 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00191371 CAS Number: 22795-99-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...