(S)-(-)-2-Acetoxypropionic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1299-25G A1299-5G
TCA1299-25GEA 591.1 USD
TCA1299-25G TCA1299-005G
(S)-(-)-2-Acetoxypropionic acid ≥98.0% (by titrimetric analysis)
(S)-(-)-2-Acetoxypropionic acid

CAS Number: 6034-46-4
MDL Number: MFCD00191370
Molecular Formula: C5H8O4
Molecular Weight: 132.12
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 110
Specific Gravity (20/20): 1.18
Specific rotation [a]20/D: -49.5 deg (C=7, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (-)-O-Acetyl-L-lactic Acid

Formula: C₅H₈O₄
Boiling Pt: 142 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00191370
CAS Number: 6034-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=104d17ae5ce206a8f0ae73387fe5e2bfc6d46d60&vid=104d17ae5ce206a8f0ae73387fe5e2bfc6d46d60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR