4-Ethynylpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 2510-22-7
MDL Number: MFCD04038420
Molecular Formula: C7H5N
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 98
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Pyridylacetylene
Formula:
C₇H₅N Melting Pt: 98 °C |
MDL Number:
MFCD04038420 CAS Number: 2510-22-7 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...