(S)-(-)-2-Acetoxypropionyl chloride ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-(-)-2-acetoxypropionylchloride, (S)-(-)-O-Acetyllactoyl Chloride
A1297-1G
A1297-5G
TCA1297-001GEA
44
USD
TCA1297-001G
TCA1297-5G
(S)-(-)-2-Acetoxypropionyl chloride ≥97.0% (by GC, titration analysis)
(S)-(-)-2-Acetoxypropionyl chloride
CAS Number: 36394-75-9
MDL Number: MFCD00145252
Molecular Formula: C5H7ClO3
Molecular Weight: 150.56
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 110
Specific Gravity (20/20): 1.19
Specific rotation [a]20/D: -31 deg (C=4, CHCl3)
Storage Temperature: 0-10°C
Other Product Names / Synonyms: (S)-(-)-O-Acetyllactoyl Chloride
Formula:
C₅H₇ClO₃ MW: 150.56 g/mol Boiling Pt: 78 °C Density: 1.19 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00145252 CAS Number: 36394-75-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...