2-Aminoethyl isopropyl ether ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 81731-43-3
MDL Number: MFCD00191603
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 120
Specific Gravity (20/20): 0.84
Other Product Names / Synonyms: 2-Isopropoxyethylamine
Formula:
C₅H₁₃NO Boiling Pt: 120 °C Density: 0.84 (20 °C) |
MDL Number:
MFCD00191603 CAS Number: 81731-43-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...