2-Ethoxy-5-nitropyridine ≥98.0%

Supplier: TCI America

E0536-25G E0536-5G
TCE0536-25GEA 187 USD
TCE0536-25G TCE0536-5G
2-Ethoxy-5-nitropyridine ≥98.0%
2-Ethoxy-5-nitropyridine

CAS Number: 31594-45-3
MDL Number: MFCD01646181
Molecular Formula: C7H8N2O3
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD01646181
CAS Number: 31594-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e07e95638329f06097c06d5a85f8d172d6923f52&vid=e07e95638329f06097c06d5a85f8d172d6923f52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR