Ethylene glycol monovinyl ether ≥98.0% stabilised

Supplier: TCI America

Synonyms: Vinyl Cellosolve, Ethylene glycol vinyl ether, 2-Vinyloxyethanol, Vinyl Glycol, 2-(Vinyloxy)ethan-1-ol

E0518-25ML E0518-500ML
TCE0518-025MLEA 42 USD
TCE0518-025ML TCE0518-500ML
Ethylene glycol monovinyl ether ≥98.0% stabilised
Ethylene glycol monovinyl ether

(stabilized with KOH)
CAS Number: 764-48-7
MDL Number: MFCD00192151
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 49
Specific Gravity (20/20): 0.98


Product Categories: Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Vinyl Cellosolve, Vinyl Glycol, 2-Vinyloxyethanol

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 143 °C
Density: 0.98 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00192151
CAS Number: 764-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c514f7b5c51cf7d8ec84fd9c23259f40ed54140&vid=6c514f7b5c51cf7d8ec84fd9c23259f40ed54140&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR