N-Ethylglycine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E0517-25G E0517-5G E0517-1G
TCE0517-25GEA 873 USD
TCE0517-25G TCE0517-005G TCE0517-1G
N-Ethylglycine ≥98.0% (by titrimetric analysis)
N-Ethylglycine

CAS Number: 627-01-0
MDL Number: MFCD00037794
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 190

Formula: C₄H₉NO₂
Melting Pt: 190 °C
MDL Number: MFCD00037794
CAS Number: 627-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=732866ce57c8de1efa0e6e4bc7d6f8d9feb2f18b&vid=732866ce57c8de1efa0e6e4bc7d6f8d9feb2f18b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR