Emodin ≥96.0% (by HPLC)
Supplier: TCI America
E0500-1G
E0500-100MG
TCE0500-1GEA
485
USD
TCE0500-1G
TCE0500-100MG
Emodin ≥96.0% (by HPLC)
Emodin
CAS Number: 518-82-1
MDL Number: MFCD00001207
Molecular Formula: C15H10O5
Molecular Weight: 270.24
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 253
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Others (Phytochemicals), Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 1,3,8-Trihydroxy-6-methylanthraquinone
Formula:
C₁₅H₁₀O₅ MW: 270.24 g/mol Melting Pt: 253 °C |
MDL Number:
MFCD00001207 CAS Number: 518-82-1 |
Specification Test Results
Purity/Analysis Method | >96.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...