2-Ethylthiazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0470-5G E0470-25G
TCE0470-005GEA 140 USD
TCE0470-005G TCE0470-25G
2-Ethylthiazole ≥98.0% (by GC, titration analysis)
2-Ethylthiazole

CAS Number: 15679-09-1
MDL Number: MFCD00085193
Molecular Formula: C5H7NS
Molecular Weight: 113.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 148
Specific Gravity (20/20): 1.06

Formula: C₅H₇NS
Boiling Pt: 148 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00085193
CAS Number: 15679-09-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2915d8a67c59638e413f2bd6564b49532baee0d&vid=c2915d8a67c59638e413f2bd6564b49532baee0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR