Ethyl glutaryl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Ethyl 5-chloro-5-oxopentanoate, Glutaric Acid Monoethyl Ester Chloride, Ethyl 4-(Chloroformyl)butyrate

E0451-5G E0451-25G
TCE0451-005GEA 36 USD
TCE0451-005G TCE0451-025G
Ethyl glutaryl chloride ≥98.0% (by titrimetric analysis)
Ethyl glutaryl chloride

CAS Number: 5205-39-0
MDL Number: MFCD00013659
Molecular Formula: C7H11ClO3
Molecular Weight: 178.61
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 82
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: Ethyl 4-(Chloroformyl)butyrate, Glutaric Acid Monoethyl Ester Chloride

Formula: C₇H₁₁ClO₃
MW: 178.61 g/mol
Boiling Pt: 108 °C
Density: 1.14 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00013659
CAS Number: 5205-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c846d66dbd624617f2970ecd08c6429428df01c6&vid=c846d66dbd624617f2970ecd08c6429428df01c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR