all cis-5,8,11,14,17-Eicosapentaenoic acid ≥95.0%

Supplier: TCI America

Synonyms: 5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic Acid, Timnodonic acid, cis-5,8,11,14,17-Eicosapentaenoic acid, EPA

E0441-100MG E0441-500MG
TCE0441-100MGEA 250 USD
TCE0441-100MG TCE0441-500MG
all cis-5,8,11,14,17-Eicosapentaenoic acid ≥95.0%
all cis-5,8,11,14,17-Eicosapentaenoic acid

CAS Number: 10417-94-4
MDL Number: MFCD00065716
Molecular Formula: C20H30O2
Molecular Weight: 302.46
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 93
Specific Gravity (20/20): 0.94
Storage Temperature: <0°C


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: EPA, all cis-5,8,11,14,17-Icosapentaenoic Acid

Formula: C₂₀H₃₀O₂
MW: 302.46 g/mol
Density: 0.94 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00065716
CAS Number: 10417-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adc65811ad40fa93e901ec7b967704b00b8d8f66&vid=adc65811ad40fa93e901ec7b967704b00b8d8f66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR