2-Amino-5-methoxybenzenesulfonic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
A1285-500G
A1285-25G
TCA1285-500GEA
652
USD
TCA1285-500G
TCA1285-025G
2-Amino-5-methoxybenzenesulfonic acid ≥97.0% (by titrimetric analysis)
2-Amino-5-methoxybenzenesulphonic acid
CAS Number: 13244-33-2
MDL Number: MFCD00035804
Molecular Formula: C7H9NO4S
Molecular Weight: 203.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Other Product Names / Synonyms: 4-Aminoanisole-3-sulfonic Acid, 2-Amino-5-methoxybenzenesulfonic Acid, 4-Methoxyaniline-2-sulfonic Acid
Formula:
C₇H₉NO₄S MW: 203.2157 g/mol |
MDL Number:
MFCD00035804 CAS Number: 13244-33-2 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...