N-(2-Methoxyethyl)ethylamine ≥97.0%

Supplier: TCI America

Synonyms: N-Ethyl-2-methoxyethanamine, N-Ethyl-2-methoxyethylamine

E0397-25G E0397-5G
TCE0397-25GEA 335 USD
TCE0397-25G TCE0397-005G
N-(2-Methoxyethyl)ethylamine ≥97.0%
N-(2-Methoxyethyl)ethylamine

CAS Number: 34322-82-2
MDL Number: MFCD00144876
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: N-Ethyl-2-methoxyethylamine

Formula: C₅H₁₃NO
Boiling Pt: 116 °C
Density: 0.83 (20 °C)
MDL Number: MFCD00144876
CAS Number: 34322-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b446a84b6fa730eb905c96c7d170e57fc52eac0&vid=5b446a84b6fa730eb905c96c7d170e57fc52eac0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR