Etilefrine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Ethylamino-1-(3-hydroxyphenyl)ethanol Hydrochloride
E0381-25G
E0381-5G
TCE0381-025GEA
355
USD
TCE0381-025G
TCE0381-5G
Etilefrine hydrochloride ≥98.0% (by HPLC, titration analysis)
Etilefrine hydrochloride
CAS Number: 943-17-9
MDL Number: MFCD00060167
Molecular Formula: C10H15NO2
Molecular Weight: 217.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 122
Product Categories: Bioscience, Other Pharmaceutical Ingredients, Phenylethylamines
Other Product Names / Synonyms: 2-Ethylamino-1-(3-hydroxyphenyl)ethanol Hydrochloride
Formula:
C₁₀H₁₅NO₂ MW: 217.69 g/mol Melting Pt: 122 °C |
MDL Number:
MFCD00060167 CAS Number: 943-17-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...