2-Ethylcyclohexanol ≥97.0%

Supplier: TCI America

E0365-25ML E0365-500ML
TCE0365-025MLEA 109.89 USD
TCE0365-025ML TCE0365-500ML
2-Ethylcyclohexanol ≥97.0%
2-Ethylcyclohexanol

CAS Number: 3760-20-1
MDL Number: MFCD00001445
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 68
Specific Gravity (20/20): 0.93

Formula: C₈H₁₆O
Boiling Pt: 79 °C
Density: 0.93 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00001445
CAS Number: 3760-20-1
EINECS: 223-174-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e37f5f1736c394a1abf0078e0e37c4398b20c2d&vid=1416d72dbdb85e3e4685519b5541753271903f11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR