Ethyl (ethoxymethylene)cyanoacetate ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Ethyl 2-Cyano-3-Ethoxyacrylate

E0341-500G E0341-25G
TCE0341-500GEA 223 USD
TCE0341-500G TCE0341-025G
Ethyl (ethoxymethylene)cyanoacetate ≥98.0% for synthesis
Ethyl (ethoxymethylene)cyanoacetate

CAS Number: 94-05-3
MDL Number: MFCD00009136
Molecular Formula: C8H11NO3
Molecular Weight: 169.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 191
Melting point (°C): 51
Flash Point (°C): 130


Other Product Names / Synonyms: 2-Cyano-3-ethoxyacrylic Acid Ethyl Ester, 2-(Ethoxymethylene)-2-cyanoacetic Acid Ethyl Ester, Ethyl 2-Cyano-3-ethoxyacrylate

Formula: C₈H₁₁NO₃
Boiling Pt: 191 °C
Melting Pt: 51 °C
Flash Pt: 130 °C
MDL Number: MFCD00009136
CAS Number: 94-05-3
EINECS: 202-299-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05d97a21261c6c814dd8cae84e957083495d90ad&vid=05d97a21261c6c814dd8cae84e957083495d90ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR