Ethyl vinyl sulfide ≥93.0%
Supplier: TCI America
E0335-5ML
E0335-25ML
TCE0335-005MLEA
107
USD
TCE0335-005ML
TCE0335-25ML
Ethyl vinyl sulfide ≥93.0%
Ethyl vinyl sulphide
CAS Number: 627-50-9
MDL Number: MFCD00015180
Molecular Formula: C4H8S
Molecular Weight: 88.17
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): -6
Specific Gravity (20/20): 0.88
Formula:
C₄H₈S Boiling Pt: 91 °C Density: 0.88 (20 °C) Flash Pt: -6 °C |
MDL Number:
MFCD00015180 CAS Number: 627-50-9 |
Specification Test Results
Purity/Analysis Method | >93.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...