Ethylene glycol monoisobutyl ether ≥98.0%

Supplier: TCI America

Synonyms: Isobutyl Cellosolve, 2-Isobutoxyethanol

E0301-500ML E0301-25ML
TCE0301-500MLEA 81 USD
TCE0301-500ML TCE0301-025ML
Ethylene glycol monoisobutyl ether ≥98.0%
Ethylene glycol monoisobutyl ether

CAS Number: 4439-24-1
MDL Number: MFCD00059601
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 57
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: 2-Isobutoxyethanol, Isobutyl Cellosolve

Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 160 °C
Density: 0.89 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00059601
CAS Number: 4439-24-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d3eaa4e40f4c736021b5caabd07261576fe3de3&vid=1d3eaa4e40f4c736021b5caabd07261576fe3de3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR