3-(2-Ethylhexyloxy)propylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Aminopropyl 2-Ethylhexyl ether, n-Propanolamine 2-Ethylhexyl ether, 3-Aminopropyl Octyl ether, 3-((2-Ethylhexyl)oxy)propan-1-amine

E0295-25ML E0295-500ML
TCE0295-025MLEA 19 USD
TCE0295-025ML TCE0295-500ML
3-(2-Ethylhexyloxy)propylamine ≥98.0% (by GC, titration analysis)
3-(2-Ethylhexyloxy)propylamine

CAS Number: 5397-31-9
MDL Number: MFCD00025619
Molecular Formula: C11H25NO
Molecular Weight: 187.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 103
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: 3-Aminopropyl 2-Ethylhexyl Ether, 3-Aminopropyl Octyl Ether, n-Propanolamine 2-Ethylhexyl Ether

Formula: C₁₁H₂₅NO
MW: 187.3223 g/mol
Boiling Pt: 78 °C
Density: 0.85 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00025619
CAS Number: 5397-31-9
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51645986093abbcd58af17727b4f39d208958316&vid=51645986093abbcd58af17727b4f39d208958316&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR