Ethoxyquin ≥90.0%
Supplier: TCI America
Synonyms:
6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, Ethoxyquin
[Antioxidant for rubber]
CAS Number: 91-53-2
MDL Number: MFCD00023883
Molecular Formula: C14H19NO
Molecular Weight: 217.31
Purity/Analysis Method: >90.0% (GC)
Form: Liquid
Boiling point (°C): 161
Flash Point (°C): 137
Specific Gravity (20/20): 1.04
Product Categories: Materials Chemistry, Resin Additives, Bioscience, Antioxidants
Other Product Names / Synonyms: 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, Ethoxyquin
Formula:
C₁₄H₁₉NO Boiling Pt: 161 °C Density: 1.04 (20 °C) Flash Pt: 137 °C |
MDL Number:
MFCD00023883 CAS Number: 91-53-2 EINECS: 202-075-7 |
|
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...