Ethyl phosphate (mono and di ester mixture)

Supplier: TCI America

E0236-500ML E0236-25ML
TCE0236-500MLEA 117.2 USD
TCE0236-500ML TCE0236-025ML
Ethyl phosphate (mono and di ester mixture)
Ethyl phosphate (mono and di ester mixture)

CAS Number: 37203-76-2
MDL Number: MFCD00136002
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 193
Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: Phosphoric Acid Ethyl Ester

Density: 1.28 (20 °C)
Flash Pt: 193 °C
MDL Number: MFCD00136002
CAS Number: 37203-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83f86e87d4710947354a38120d4145d09507e053&vid=83f86e87d4710947354a38120d4145d09507e053&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR