N-Ethyl-N-2-hydroxyethyl-m-toluidine ≥96.0%

Supplier: TCI America

Synonyms: 2-(Ethyl(m-tolyl)amino)ethanol, 2-[Ethyl(3-methylphenyl)amino]ethan-1-ol

E0220-25G E0220-500G
TCE0220-025GEA 25 USD
TCE0220-025G TCE0220-500G
N-Ethyl-N-2-hydroxyethyl-m-toluidine ≥96.0%
N-Ethyl-N-2-hydroxyethyl-m-toluidine

CAS Number: 91-88-3
MDL Number: MFCD00002849
Molecular Formula: C11H17NO
Molecular Weight: 179.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 154
Specific Gravity (20/20): 1.02

Formula: C₁₁H₁₇NO
MW: 179.26 g/mol
Boiling Pt: 147 °C
Density: 1.02 (20 °C)
Flash Pt: 154 °C
MDL Number: MFCD00002849
CAS Number: 91-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=141be8d8f733d50538ff82d5fe5e10436004bde0&vid=141be8d8f733d50538ff82d5fe5e10436004bde0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR