N-Ethyltoluenesulfonamide(o and p mixture) ≥98.0% (by total nitrogen basis)
Supplier: TCI America
E0187-25G
E0187-500G
TCE0187-025GEA
28
USD
TCE0187-025G
TCE0187-500G
N-Ethyltoluenesulfonamide(o and p mixture) ≥98.0% (by total nitrogen basis)
N-Ethyltoluenesulfonamide(o and p mixture)
CAS Number: 76902-32-4
MDL Number: MFCD00048511
Molecular Formula: C9H13NO2S
Molecular Weight: 199.27
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Flash Point (°C): 193
Specific Gravity (20/20): 1.21
Formula:
C₉H₁₃NO₂S Density: 1.21 (20 °C) Flash Pt: 193 °C |
MDL Number:
MFCD00048511 CAS Number: 76902-32-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...