S-Ethylisothiouronium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Ethyl carbamimidothioate hydrobromide, S-Ethylisothiourea Hydrobromide, S-EIT hydrobromide, 2-Ethyl-2-thiopseudourea hydrobromide

E0182-25G E0182-100G
TCE0182-025GEA 188 USD
TCE0182-025G TCE0182-100G
S-Ethylisothiouronium bromide ≥98.0% (by titrimetric analysis)
S-Ethylisothiouronium bromide

CAS Number: 1071-37-0
MDL Number: MFCD00012585
Molecular Formula: C3H8N2S
Molecular Weight: 185.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 90

Formula: C₃H₉BrN₂S
MW: 185.1 g/mol
Melting Pt: 90 °C
MDL Number: MFCD00012585
CAS Number: 1071-37-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4576cba4c4c221a045dae6bfae089f6c3b83ad97&vid=4576cba4c4c221a045dae6bfae089f6c3b83ad97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR