Ethylenediaminetetraacetic acid lead(II) disodium salt hydrate ≥98.0%

Supplier: TCI America

Synonyms: Lead(II) Disodium Ethylenediaminetetraacetate

E0093-500G E0093-25G
TCE0093-500GEA 559.7 USD
TCE0093-500G TCE0093-025G
Ethylenediaminetetraacetic acid lead(II) disodium salt hydrate ≥98.0%
Ethylenediaminetetraacetic acid lead(II) disodium salt hydrate

CAS Number: 22904-40-1
MDL Number: MFCD00148921
Molecular Formula: C10H16N2O8
Molecular Weight: 541.39
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Lead(II) Disodium Ethylenediaminetetraacetate

Formula: C₁₀H₁₆N₂O₈
MW: 541.39 g/mol
MDL Number: MFCD00148921
CAS Number: 22904-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8985d91ff2c9b7bd5b67bccb17b70a0b9777ed80&vid=8985d91ff2c9b7bd5b67bccb17b70a0b9777ed80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR